Skip to Content
Merck
All Photos(1)

Documents

330469

Sigma-Aldrich

N-Phenylglycine

97%

Synonym(s):

(Phenylamino)acetic acid, Anilinoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH2COOH
CAS Number:
Molecular Weight:
151.16
Beilstein:
509838
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

mp

121-123 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CNc1ccccc1

InChI

1S/C8H9NO2/c10-8(11)6-9-7-4-2-1-3-5-7/h1-5,9H,6H2,(H,10,11)

InChI key

NPKSPKHJBVJUKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M Janusz et al.
Journal of medicinal chemistry, 33(3), 1052-1061 (1990-03-01)
Twenty esters of L-aspartyl-D-phenylglycine, as well as two substituted analogues, an o-fluoro and a p-hydroxy-phenylglycine ester, were prepared. The L-aspartyl-D-phenylglycine (-)-alpha- and (+)-beta-fenchyl esters had the highest sweetness potency at 1200 and 3700 times that of sucrose, respectively. The high
G E Schumacher et al.
Journal of dental research, 76(1), 602-609 (1997-01-01)
Effective composite-to-dentin bonding has been achieved by the sequential use of dilute aqueous nitric acid (HNO3) and acetone solutions of N-phenylglycine and a carboxylic acid monomer, e.g., p-PMDM. Both the HNO3 pre-treatment and the surface-initiated polymerization that results from reaction
R E Webb et al.
Journal of dental research, 70(3), 211-214 (1991-03-01)
Three structurally related substituted amino acids (N-compounds) were studied in a three-step dentin-bonding protocol. The first step of an acidic ferric oxalate solution and the third step of a surface-active comonomer were held constant throughout the study. In the second
G E Schumacher et al.
Dental materials : official publication of the Academy of Dental Materials, 8(4), 278-282 (1992-07-01)
Effective dentin bonding systems based on para-PMDM diadduct of pyromellitic dianhydride and 2-hydroxyethyl methacrylate (HEMA) have been developed (Bowen et al., 1982). Para-PMDM, a solid of limited solubility, is usually applied from an acetone solution to dentin that has been
Y Imai et al.
Dental materials : official publication of the Academy of Dental Materials, 10(4), 275-277 (1994-07-01)
This research was designed to study the effect of water or carboxylic acid monomer on the polymerization of 2-hydroxyethyl methacrylate (HEMA) in order to understand the bonding mechanism of dentin bonding systems using N-phenylglycine (NPG). The polymerization of HEMA in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service