Skip to Content
Merck
All Photos(1)

Key Documents

326526

Sigma-Aldrich

Gold

wire, diam. 0.127 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

wire

resistivity

2.05 μΩ-cm, 0°C

diam.

0.127 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Quantity

500 mg = 2 m; 2.5 g = 10 m

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengxia Liu et al.
Journal of biomedical nanotechnology, 9(6), 1017-1028 (2013-07-19)
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on
Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the
Tianxun Gong et al.
Journal of biomedical nanotechnology, 9(6), 985-991 (2013-07-19)
In this paper, we proposed the use of gold nanoparticles as plasmon scattering probes for dark-field multiplex imaging of live cancer cells. By carefully engineering the surfaces, aqueous dispersions of anti-EGFR antibody-conjugated gold nanospheres and gold nanorods are prepared. We
Claudia Diletto et al.
Journal of nanoscience and nanotechnology, 13(7), 5215-5220 (2013-08-02)
Innovative hybrid nanocomposites based on a nanostructured block copolymer (BCP) matrix whose lamellar nanodomains are selectively loaded with metal nanoparticles, have been prepared. A symmetric poly(styrene-b-methylmethacrylate) (PS-b-PMMA) amorphous BCP showing a lamellar morphology has been employed. Thin films of PS-b-PMMA
Fengxian Zheng et al.
Journal of nanoscience and nanotechnology, 13(6), 3990-3998 (2013-07-19)
Gold nanoparticles (AuNPs) are of biomedical importance, such as delivery vectors. Therefore, we used all-atom molecular dynamic simulations to study the interaction of AuNPs with cell membrane (DMPC bilayer). We observed that the AuNPs adhered spontaneously on the surface of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service