Skip to Content
Merck
All Photos(3)

Documents

325104

Sigma-Aldrich

3-Nitrophenylboronic acid

≥97%

Synonym(s):

3-Nitrobenzeneboronic acid, m-Nitrobenzeneboronic acid, m-Nitrophenylboronic acid, NSC 401539, NSC 59739

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4B(OH)2
CAS Number:
Molecular Weight:
166.93
Beilstein:
2938638
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder

mp

284-285 °C (dec.) (lit.)

SMILES string

OB(O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C6H6BNO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4,9-10H

InChI key

ZNRGSYUVFVNSAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Copper-catalyzed arylation
  • Palladium-catalyzed decarboxylative coupling
  • Suzuki-Miyaura cross-coupling
  • Oxidative carbocyclization / arylation
  • Addition to arylpropargyl alcohols

Additionally used as a reactant for synthesizing biologically active molecules such as:
  • Inhibitors of angiogenesis
  • Biaryl-olefins with antiproliferative activities
Catalyzes ene carbocyclization of acetylenic dicarbonyl compounds

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meiling Li et al.
Chemical communications (Cambridge, England), 46(13), 2191-2193 (2010-03-18)
The discovery and development of an efficient ene carbocyclization of 1,3-dicarbonyl compounds bearing pendent terminal alkyne substituents under 3-nitrobenzeneboronic acid catalysis is described. The reaction is efficient, easy to perform and general to a wide range of ketoester substrates.
Chiaki Miyamoto et al.
Inorganic chemistry, 47(5), 1417-1419 (2008-02-12)
The rate constants for a boronate ion were determined for the first time using the reaction systems of 3-nitrophenylboronic acid (3-NO2PhB(OH)2) with ethylene glycol (EG) and propylene glycol (PG) in an alkaline solution: the rate constants (25 degrees C, I
Maria T Gallardo-Williams et al.
The Prostate, 54(1), 44-49 (2002-12-14)
Prostate specific antigen (PSA) is a well-established marker of prostate cancer, but it can also degrade extracellular matrix proteins such as fibronectin and could be involved in tumor progression and metastasis. In this study, we have addressed the use of
Chun-Ping Lu et al.
Journal of agricultural and food chemistry, 59(21), 11403-11406 (2011-10-05)
Hydrogen peroxide is commonly used in the food processing industry as a chlorine-free bleaching and sterilizing agent, but excessive amounts of residual hydrogen peroxide have led to cases of food poisoning. Here we describe the development of a novel nonenzymatic
Wei Ke et al.
Antimicrobial agents and chemotherapy, 56(5), 2713-2718 (2012-02-15)
Class A carbapenemases are a major threat to the potency of carbapenem antibiotics. A widespread carbapenemase, KPC-2, is not easily inhibited by β-lactamase inhibitors (i.e., clavulanic acid, sulbactam, and tazobactam). To explore different mechanisms of inhibition of KPC-2, we determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service