Skip to Content
Merck
All Photos(1)

Documents

294357

Sigma-Aldrich

DL-Homophenylalanine

98%

Synonym(s):

(±)-2-Amino-4-phenylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2CH2)CH(NH2)CO2H
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

282 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(CCc1ccccc1)C(O)=O

InChI

1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)

InChI key

JTTHKOPSMAVJFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adeyemi O Aremu et al.
Journal of plant physiology, 169(15), 1530-1541 (2012-08-14)
The effect of five topolins (meta-Topolin=mT; meta-Topolin riboside=mTR; meta-Methoxy topolin=MemT; meta-Methoxy topolin riboside=MemTR and 6-(meta-methoxy)-9-(tetrahydropyran-2-yl)-topolin=MemTTHP) on the photosynthetic pigments and leaf structures of micropropagated 'Williams' bananas was compared with the commonly used benzyladenine (BA). Surface-decontaminated explants were cultured for 70
S R Syam Prakash et al.
Journal of plant physiology, 163(11), 1135-1149 (2006-05-24)
Calcium-dependent protein kinases (CPKs) play important roles in multiple signal transduction pathways but the precise role of individual CPK is largely unknown. We isolated two cDNAs encoding two CPK isoforms (Cicer arietinum CPKs-CaCPK1 and CaCPK2) of chickpea. Their expression in
Daniele Balducci et al.
Amino acids, 38(3), 829-837 (2009-04-14)
A new and convenient stereocontrolled synthesis of the optically pure (S)-alpha-methyl,alpha-amino acids 6(a-d) that exploits the chiral synthon 1,4-N,N-[(S)-1-phenylethyl]-piperazine-2,5-dione (1) is described. The (S)-1-phenylethyl group, bonded to each of the N-atoms of the 2,5-diketopiperazine, acts as a chiral inductor in
A L Ahmad et al.
Biotechnology advances, 27(3), 286-296 (2009-06-09)
Over the past decade, L-homophenylalanine is extensively used in the pharmaceutical industry as a precursor for production of angiotensin-converting enzyme (ACE) inhibitor, which possesses significant clinical application in the management of hypertension and congestive heart failure (CHF). A number of
Chao-Hung Kao et al.
Journal of biotechnology, 134(3-4), 231-239 (2008-03-18)
A dihydropyrimidinase gene (pydB) was cloned from the moderate thermophilic Brevibacillus agri NCHU1002 and expressed in Escherichia coli. The purified dihydropyrimidinase exhibited strict d-enantioselectivity for D,L-p-hydroxyphenylhydantoin and D,L-5-[2-(methylthio)ethyl]hydantoin, and non-enantiospecificity for D,L-homophenylalanylhydantoin (D,L-HPAH). The hydrolytic activity of PydB was enhanced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service