Skip to Content
Merck
All Photos(1)

Key Documents

291463

Sigma-Aldrich

Glycidyl isopropyl ether

98%

Synonym(s):

2,3-Epoxypropyl isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
Beilstein:
103028
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

refractive index

n20/D 1.410 (lit.)

bp

131-132 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

SMILES string

CC(C)OCC1CO1

InChI

1S/C6H12O2/c1-5(2)7-3-6-4-8-6/h5-6H,3-4H2,1-2H3

InChI key

NWLUZGJDEZBBRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Ting Chang et al.
Bioresource technology, 97(11), 1265-1271 (2005-07-26)
China fir (Cunninghamia lanceolata var. lanceolata) and maple (Acer sp.) wood were etherified with isopropyl glycidyl ether and the decay resistance and light stability of the modified wood were assessed. CP/MAS (13)C NMR and FT-IR analyses indicated that new ether
P Foureman et al.
Environmental and molecular mutagenesis, 23(1), 51-63 (1994-01-01)
Fifty chemicals were tested for mutagenic activity in post-meiotic and meiotic germ cells of male Drosophila melanogaster using the sex-linked recessive lethal (SLRL) assay. As in the previous studies in this series, feeding was chosen as the first route of
R Zeeman et al.
Journal of biomedical materials research, 51(4), 541-548 (2000-07-06)
Dermal sheep collagen was crosslinked with 1,4-butanediol diglycidyl ether (BDDGE) or modified with glycidyl isopropyl ether (PGE). The reduction in amine groups as a function of time was followed to study the overall reaction kinetics of collagen with either BDDGE
Takashi Toyao et al.
ChemSusChem, 8(22), 3905-3912 (2015-09-24)
Various N-doped nanoporous carbons containing metal species were prepared by direct thermal conversion of zeolitic imidazolate frameworks (ZIFs; ZIF-7, -8, -9, and -67) at different temperatures (600, 800, and 1000 °C). These materials were utilized as bifunctional acid-base catalysts to promote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service