Skip to Content
Merck
All Photos(1)

Key Documents

278041

Sigma-Aldrich

Diethyl tert-butylmalonate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH(COOC2H5)2
CAS Number:
Molecular Weight:
216.27
Beilstein:
1781709
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

102-104 °C/11 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C(=O)OCC)C(C)(C)C

InChI

1S/C11H20O4/c1-6-14-9(12)8(11(3,4)5)10(13)15-7-2/h8H,6-7H2,1-5H3

InChI key

RJNICNBRGVKNSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl tert-butylmalonate has been used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide (TBIPPS).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X L Ju et al.
Bioorganic & medicinal chemistry, 8(9), 2337-2341 (2000-10-12)
The enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2 ]octane 1-sulfide (TBIPPS) were prepared in nine steps from diethyl tert-butylmalonate, and their abilities to compete with [3H]1-(4-ethynylphenyl)-4-n-propyl-2,6,7-trioxabicyclo[2.2.2 ]octane (EBOB), a noncompetitive antagonist of ionotropic gamma-aminobutyric acid (GABA) receptors, at their binding site were investigated using
Grazia Luisi et al.
Antioxidants (Basel, Switzerland), 8(1) (2018-12-28)
Oxidative damage is among the factors associated with the onset of chronic pathologies, such as neurodegenerative and metabolic diseases. Several classes of anti-oxidant compounds have been suggested as having a protective role against cellular stressors, but, in this perspective, peptides'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service