Skip to Content
Merck
All Photos(1)

Key Documents

265020

Sigma-Aldrich

Dimethylphenylphosphine

99%

Synonym(s):

Me2PPh, Phenyldimethylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2PC6H5
CAS Number:
Molecular Weight:
138.15
Beilstein:
742064
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

11 mmHg ( 68 °C)

Assay

99%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.563 (lit.)

bp

74-75 °C/12 mmHg (lit.)

density

0.971 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CP(C)c1ccccc1

InChI

1S/C8H11P/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

HASCQPSFPAKVEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Yang et al.
Colloids and surfaces. B, Biointerfaces, 160, 136-143 (2017-09-19)
Hydroxyapatite-based implant is widely used in the fields of artificial teeth and bones. However, the problem of microbial communities on hydroxyapatite surfaces remain a great challenge. In order to inhibit bacterial adhesion and activity in a long term, a salivary
Mohammad Mahdi Bordbar et al.
Scientific reports, 10(1), 17302-17302 (2020-10-16)
This study introduces an applicable colorimetric sensor array for the detection of pesticides in the vapor phase. The array consisted of six metal nanoparticles spotted on the piece of filter paper. 3D-origami pattern was used for the fabrication of a
Fan Zhang et al.
Journal of the American Chemical Society, 140(42), 13854-13868 (2018-10-03)
Inorganic oxides play a crucial role in the activation of atomically dispersed metal oxides for catalytic olefin transformations, but the inefficient activation processes remain poorly understood. Activation of methyltrioxorhenium (MTO) for propene metathesis via its deposition on the surface of
Omed Al-Jaf et al.
Soft matter, 13(10), 2075-2084 (2017-02-22)
The nanomechanical properties of zwitterionic poly(cysteine methacrylate) (PCysMA) brushes grown from planar surfaces by atom transfer radical polymerisation have been characterised by friction force microscopy (FFM). FFM provides quantitative insights into polymer structure-property relationships and in particular illuminates the dependence
Joseph S Brown et al.
Organic & biomolecular chemistry, 18(32), 6364-6377 (2020-08-08)
The thiol-Michael addition is a popular, selective, high-yield "click" reaction utilized for applications ranging from small-molecule synthesis to polymer or surface modification. Here, we combined experimental and quantum mechanical modeling approaches using density functional theory (DFT) to examine the thiol-Michael

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service