Skip to Content
Merck
All Photos(2)

Documents

204951

Sigma-Aldrich

Zinc oxide

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
Molecular Weight:
81.39
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38150402
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ZnO is a semiconductor with a wide band gap. Point defects in the structure define the electrical conductivity of the oxide. Doping these point defects with Al, In and Ga gives in a highly conductive n –type zinc oxide. Defect free ZnO can also be achieved by annealing the oxide in reducing environment. It also exhibits excellent optical transmission properties. Doped ZnO can form transparent conductors which may be useful in various energy-based applications.1

Application

Zinc oxide was added to the diet of piglets to enhance their growth performance and health.
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Zinc Concentration in the Diet and the Length of the Feeding Period Affect the Methylation Status of the ZIP4 Zinc Transporter Gene in Piglets
Karweina D, et al.
PLoS ONE, 10(11), e0143098-e0143098 (2015)
A. M. Healey et al.
Inorganic chemistry, 38(3), 455-458 (2001-10-25)
The structure of NaZnSiO(3)OH, synthesized hydrothermally by reaction of Na(2)ZnSiO(4) and NaOH, has been determined from single-crystal X-ray and powder neutron diffraction data (orthorhombic, space group P2(1)2(1)2(1,) a = 7.6872(2) Å, b = 9.3899(2) Å, c = 5.155(1) Å, Z
Hyeong Pil Kim et al.
Journal of nanoscience and nanotechnology, 13(7), 5142-5147 (2013-08-02)
Solution processed cathode organic photovoltaic cells (OPVs) utilizing thin layer of ZnO with 27% increase in power conversion efficiency (PCE) to control devices have been demonstrated. Devices without the presence of ZnO layer have much lower PCE than the ones
Jeongmin Kang et al.
Journal of nanoscience and nanotechnology, 13(5), 3526-3528 (2013-07-19)
ZnO-nanowire-based logic circuits were constructed by the vertical integration of multilayered field-effect transistors (FETs) on plastic substrates. ZnO nanowires with an average diameter of -100 nm were synthesized by thermal chemical vapor deposition for use as the channel material in
Alexander Lange et al.
Journal of nanoscience and nanotechnology, 13(7), 5209-5214 (2013-08-02)
Organic solar cells are a favorable alternative to their inorganic counterparts because the functional layers of these devices can be processed with printing or coating on a large scale. In this study, a novel polymer was synthesized, blended with fullerene

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service