17988
DL-β-Leucine
≥98.0% (NT)
Synonym(s):
(±)-3-Amino-4-methylpentanoic acid, (±)-3-Amino-4-methylvaleric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98.0% (NT)
application(s)
peptide synthesis
SMILES string
CC(C)C(N)CC(O)=O
InChI
1S/C6H13NO2/c1-4(2)5(7)3-6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)
InChI key
GLUJNGJDHCTUJY-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of biological chemistry, 255(21), 10067-10072 (1980-11-10)
Circulating levels of beta-leucine are elevated in the cobalamin-deficient state of pernicious anemia. Levels of leucine, on the other hand, are much lower. It is proposed that leucine 2,3-aminomutase, the cobalamin-dependent enzyme that catalyzes the interconversion of leucine and beta-leucine
Cobalamin-dependent leucine and beta-leucine synthesis in higher animals.
Nutrition reviews, 39(6), 244-246 (1981-06-01)
The Journal of nutrition, 118(2), 159-164 (1988-02-01)
This study was designed to investigate a postulated relationship between vitamin B-12 and leucine metabolism in mature domestic chickens. Plasma amino acid analysis revealed the presence of beta-leucine at a concentration of 60 to 80 mumol/l. After 425 d on
Beta-leucine and the beta-keto pathway of leucine metabolism.
Advances in enzymology and related areas of molecular biology, 58, 173-189 (1986-01-01)
The synthesis and detection of beta leucines.
Proceedings of the Western Pharmacology Society, 24, 117-117 (1981-01-01)
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service