Skip to Content
Merck
All Photos(2)

Documents

158410

Sigma-Aldrich

5-Bromovaleric acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)4COOH
CAS Number:
Molecular Weight:
181.03
Beilstein:
1745664
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

38-40 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear, colorless to faintly yellow

SMILES string

OC(=O)CCCCBr

InChI

1S/C5H9BrO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)

InChI key

WNXNUPJZWYOKMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromovaleric acid was used in the preparation of:
  • 5-(3-dimethylammonio)propylammonio pentanoate bromide via reaction with 1,1-dimethyl-1,3-propylenediamine
  • nanodiamond functionalized by carboxylic acid groups
  • water-soluble nanoplatelets of potassium graphite

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleksandr Kuznetsov et al.
Langmuir : the ACS journal of surfaces and colloids, 28(11), 5243-5248 (2012-03-01)
Reduction of the graphenic edges of annealed nanodiamond by sodium in liquid ammonia leads to a nanodiamond salt that reacts with either alkyl or aryl halides by electron transfer to yield radical anions that dissociate spontaneously into free radicals and
Natashya Falcone et al.
Chembiochem : a European journal of chemical biology, 20(6), 838-845 (2018-12-01)
Nicotinamide and pyridine-containing conjugates have attracted a lot of attention in research as they have found use in a wide range of applications including as redox flow batteries and calcium channel blockers, in biocatalysis, and in metabolism. The interesting redox
Functionalization of potassium graphite.
Soma Chakraborty et al.
Angewandte Chemie (International ed. in English), 46(24), 4486-4488 (2007-05-05)
Na Peng et al.
Acta biomaterialia, 105, 223-238 (2020-01-12)
Targeting delivery of photosensitizers to mitochondria as the most sensitive cellular organelles to reactive oxygen species (ROS) by positively charged polymeric nanocarriers (NCs) is one of the useful methods for efficient photodynamic therapy (PDT). However, the NCs with positively charged
Caixia Yue et al.
Theranostics, 6(13), 2352-2366 (2016-11-24)
Mitochondria in cancer cells maintain a more negative membrane potential than normal cells. Mitochondria are the primary source of cellular reactive oxygen species (ROS), which are necessary for photodynamic therapy. Thus, the strategy of targeting mitochondria can maximize the photodynamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service