Skip to Content
Merck
All Photos(1)

Key Documents

13371

Sigma-Aldrich

Benzyldimethyldecylammonium chloride

≥97.0% (AT)

Synonym(s):

Benzyldecyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
311.93
Beilstein:
3578562
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (AT)

form

powder

mp

50-60 °C

SMILES string

[Cl-].CCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzyldimethyldecylammonium chloride, also known as BAC-C10, is commonly used as a cationic surfactant.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrocoagulation-electrooxidation for mitigating trace organic compounds in model drinking water sources
Ryan DR, et al.
Chemosphere, 273, 129377-129377 (2021)
Adsorptive removal of cationic surfactants from aqueous solutions onto high-area activated carbon cloth monitored by in situ UV spectroscopy
Duman O and Ayranci E
Journal of Hazardous Materials, 174(1-3), 359-367 (2010)
Niels van Boxtel et al.
Journal of chromatography. A, 1476, 105-113 (2016-11-26)
The analysis of quaternary ammonium salts (QAS) using GC is often performed by "in injector" pyrolysis to create volatile degradation products for quantification purposes. Besides the risk of severe system contamination, the application of this approach on aqueous samples is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service