119334
Bis(cyclopentadienyl)hafnium(IV) dichloride
98%
Synonym(s):
Di(cyclopentadienyl)hafnium(IV) dichloride, Hafnocene dichloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H10Cl2Hf
CAS Number:
Molecular Weight:
379.58
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
reaction suitability
core: hafnium
reagent type: catalyst
mp
230-233 °C (lit.)
SMILES string
Cl[Hf]Cl.[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2
InChI
1S/2C5H5.2ClH.Hf/c2*1-2-4-5-3-1;;;/h2*1-5H;2*1H;/q;;;;+2/p-2
InChI key
UHCOEQZUSAFLHQ-UHFFFAOYSA-L
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Köpf-Maier et al.
Journal of cancer research and clinical oncology, 96(1), 43-51 (1980-01-01)
The antitumor activity of the metallocene dichlorides TDC, ZDC, and HDC against Ehrlich ascites tumor in CF1 mice is investigated. A single i.p. injection of TDC in the optimum dose (30-60 mg/kg) 24 h p.t.t. achieves survival of 80-90% of
P Köpf-Maier et al.
Cancer chemotherapy and pharmacology, 5(4), 237-241 (1981-01-01)
The in vitro growth-inhibiting potencies of titanocene dichloride (TDC), zirconocene dichloride (ZDC), hafnocene dichloride (HDC), vanadocene dichloride (VDC), and molybdocene dichloride (MDC) against Ehrlich ascites tumor (EAT) cells cultured in viro as permanently growing suspension cultures were determined. The most
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service