Skip to Content
Merck
All Photos(3)

Documents

116068

Sigma-Aldrich

1-(2-Hydroxyethyl)piperidine

ReagentPlus®, 99%

Synonym(s):

1-Piperidineethanol, 2-Piperidinoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
Beilstein:
103390
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.4804 (lit.)

bp

199-202 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCCCC1

InChI

1S/C7H15NO/c9-7-6-8-4-2-1-3-5-8/h9H,1-7H2

InChI key

KZTWONRVIPPDKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(2-Hydroxyethyl)piperidine (1-Piperidineethanol) has been used as the base quencher to determine the efficiency of the decomposition of photoacid generators.
Counter-ion used for investigations of skin permeation of flurbiprofen

Catalyst used for enantioselective synthesis

Amine quencher used during synthesis of photoacid generators for ArF lithography

Adsorbent for capture and release of pressurized carbon dioxide for greenhouse gas control

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Ma et al.
Journal of pharmaceutical sciences, 99(4), 1826-1837 (2009-11-07)
The aim of this work was to investigate the percutaneous absorption of flurbiprofen (FP) using counter-ions as enhancers as well as to compare their enhancing activity with penetration enhancers in vitro. The in vitro permeation studies of FP were performed
T. Asakura, et al
J. Photopolym. Sci. Technol., 18, 407-414 (2005)
K. Tomizaki, et al.,
Chemistry Letters (Jpn), 37, 516-517 (2008)
A standard addition technique to quantify photoacid generation in chemically amplified photoresist.
Chemistry of Materials, 13(11), 4154-4162 (2001)
François Lutz et al.
Journal of the American Chemical Society, 127(35), 12206-12207 (2005-09-01)
An asymmetric autocatalytic reaction has been catalyzed by a mixture of chiral and achiral beta-amino alcohols. The absolute configuration of the highly enantioenriched obtained product (>98% ee) was shown to depend not only on the absolute configuration of the chiral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service