Skip to Content
Merck
All Photos(1)

Key Documents

112100

Sigma-Aldrich

Hexylene glycol

99%

Synonym(s):

(±)-2-Methyl-2,4-pentanediol, MPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

Quality Level

vapor pressure

0.02 mmHg ( 20 °C)

Assay

99%

form

liquid

expl. lim.

7.4 %

refractive index

n20/D 1.427 (lit.)

bp

197 °C (lit.)

mp

−40 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexylene glycol is commonly used as a solvent and as an intermediate in organic synthesis. It helps in isolating chromosomes from the human cell line BALL-1.

Application


  • pH regulatory divergent point for the selective bio-oxidation of primary diols during resting cell catalysis.: This study highlights the role of Hexylene Glycol in biocatalysis, emphasizing its application in enhancing the selective oxidation of primary diols, which is pivotal for developing efficient bio-based production processes (Hua et al., 2022).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Osamu Hoshi et al.
Methods in molecular biology (Clifton, N.J.), 736, 109-115 (2011-06-11)
Methods for atomic force microscopy (AFM) imaging of human metaphase chromosomes were -introduced in the present study. Chromosomes from the lymphocytes were fixed and prepared onto glass slides as the chromosome spread, and observed in phosphate-buffered saline by dynamic mode
The OH-initiated oxidation of hexylene glycol and diacetone alcohol
Magneron, et al.
Environmental Science & Technology, 37, 4170-4181 (2003)
Asha R Johal et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 3), 268-276 (2012-02-22)
The human ABO(H) A and B blood group glycosyltransferases GTA and GTB differ by only four amino acids, yet this small dissimilarity is responsible for significant differences in biosynthesis, kinetics and structure. Like other glycosyltransferases, these two enzymes have been
Mrignayani Kotecha et al.
Magnetic resonance in chemistry : MRC, 45 Suppl 1, S221-S230 (2007-12-25)
The use of a low-power two-pulse phase modulation (TPPM) sequence is proposed for efficient (1)H radio frequency (rf) decoupling in high-resolution (13)C solid-state NMR (SSNMR) under fast MAS conditions. Decoupling efficiency for different low-power decoupling sequences such as continuous-wave (cw)
Catherine Michaux et al.
BMC structural biology, 8, 29-29 (2008-06-05)
Recently, we reported a unique approach to preserve the activity of some proteins in the presence of the denaturing agent, Sodium Dodecyl Sulfate (SDS). This was made possible by addition of the amphipathic solvent 2,4-Methyl-2-PentaneDiol (MPD), used as protecting but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service