Supelco® SmartChemicals for Titration
Section Overview
- Titration Goes Digital—Connect Your SmartChemicals to Your Titrator
- Seamless Transfer of Titration Data via RFID Tag
- Four Benefits Using SmartChemicals for Your Titration
- SmartChemicals Are Compatible With Mettler Toledo Titrators
- Demo Video: Titration goes Digital
- Supelco® SmartChemicals - Product List
Titration Goes Digital—Connect Your SmartChemicals to Your Titrator
A new titration technology, using Supelco® SmartTitrants & Supelco® SmartStandards transfers data seamlessly to the titrator. Just a brief touch on the SmartChemical RFID (Radio Frequency Identification) tag saves time, reduces errors and always ensures maximum data integrity.
Improve your Karl Fischer Titration Process for Water Determination. Learn how to avoid errors, get tips & tricks, and discover seamless data transfer. Join the on-demand webinar
Seamless transfer of titration data via RFID tag
Using SmartChemicals eliminates time-consuming steps and errors by transferring data wirelessly and instantly. Titripur® volumetric solutions, Certipur® volumetric standards and all Aquastar® Karl Fischer titrants and standards are embedded with an RFID tag on the label. All relevant data from the Certificate of Analysis are stored on the RFID tag of the SmartChemical
Just a simple touch transfers all data in seconds
All information needed for method execution, result calculation and documentation is seamlessly transferred
- Product name
- Article number
- Lot/batch no
- Concentrations
- Date of release
- Shelf life
When using standards, the following additional information is also transferred:
- Molecular weight
- Supplier name
- Compliance according to pharmacopeias and ISO
- Measurement uncertainty
Initial opening date – stored on the tag
Previously opened chemicals are also recognized, the initial opening date is stored in the RFID tag and is shown on the touchscreen. Expired chemicals are automatically locked from use.
Four Benefits Using SmartChemicals for Your Titration
- Secure data transfer—ensures complete and correct reagent & standard data transfer in the titration software
- Ease of use—intuitive operation and convenient data transfer in one touch
- Extended quality management—titer determination, shelf life, compliance data, initial opening date, release date
- Improved efficiency—fast data transfer - safes time - no manual writing and no four-eyes principle necessary
Compact Titrator
- G20S
- G10S
- V30S
- V20S
- V10S
Excellence Titrators
- T9
- T7
- T5
External SmartChemical Reader
Titripur® Volumetric Solutions Certified According to ISO 17025
Certipur® Volumetric Standards according to ISO 17034 (Certified Reference Materials)
Aquastar® Karl Fischer Titrants