HPLC Analysis of Polyethylene Glycol 200 on TSKgel® G-OLIGO-PW
CONDITIONS
column
TSKgel G-OLIGO-PW, 30 cm × 7.8 mm I.D., 6 μm particles (808031)
mobile phase
distilled water
flow rate
1 mL/min
detector
RI
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation