Merck
  • Simultaneous determination of testosterone, cortisol, and dehydroepiandrosterone in saliva by stable isotope dilution on-line in-tube solid-phase microextraction coupled with liquid chromatography-tandem mass spectrometry.
Simultaneous determination of testosterone, cortisol, and dehydroepiandrosterone in saliva by stable isotope dilution on-line in-tube solid-phase microextraction coupled with liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2012-10-16)
Hiroyuki Kataoka, Kentaro Ehara, Rie Yasuhara, Keita Saito
ABSTRACT
We have developed a simple and sensitive method for the simultaneous determination of testosterone (TES), cortisol (CRT), and dehydroepiandrosterone (DHEA) in saliva by automated online in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-tandem mass spectrometry (LC-MS/MS) using a Discovery HS F5 column. The optimum in-tube SPME conditions were 25 draw/eject cycles of 40 μL of sample at a flow rate of 200 μL/min using a Supel-Q PLOT capillary column as an extraction device. The extracted compounds were easily desorbed from the capillary by passage of the mobile phase, and no carryover was observed. The in-tube SPME LC-MS/MS method showed good linearity with correlation coefficients r ≥ 0.9998 for TES, CRT, and DHEA using their respective stable isotope-labeled internal standards. The intra-day and inter-day precisions (relative standard deviations) were below 4.9 and 8.5 % (n = 5), respectively. This method was successfully utilized to analyze TES, CRT, and DHEA in saliva samples without any other pretreatment or interference peaks, and the quantification limits (S/N = 10) of TES, CRT and DHEA were about 0.01, 0.03 and 0.29 ng/mL saliva, respectively. The recoveries of these compounds spiked into saliva samples were each above 94%. This method was applied to analyze changes in salivary TES, CRT, and DHEA levels resulting from stress and fatigue load.
Product Description
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 1 kit
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm, pkg of 1 ea
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS F5 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS F5 HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 2.1 mm