Merck
  • Simultaneous LC-MS-MS Determination of Lopinavir and Rifabutin in Human Plasma.
Simultaneous LC-MS-MS Determination of Lopinavir and Rifabutin in Human Plasma.
Journal of chromatographic science (2017-03-24)
Swati Jaiswal, Abhisheak Sharma, Mahendra Shukla, Jawahar Lal
ABSTRACT
Tuberculosis (TB) with human immunodeficiency virus (HIV)/acquired immunodeficiency syndrome represents the most common infectious diseases worldwide. Anti-TB drugs are used concurrently with antiretroviral drug for treatment of TB-HIV co-morbidities. Due to lower risk of interaction with protease inhibitors, rifabutin is preferred over rifampicin in treatment of HIV and TB co-morbidity. A simple and specific liquid chromatography tandem mass spectrometry method was developed for quantification of rifabutin (RBT) and lopinavir (LPV) simultaneously in human plasma. Following extraction using 60% n-hexane in ethyl acetate, the processed samples were chromatographed on a Discovery HS C18 column (5 μm, 50 × 4.6 mm, id) using mobile phase [85% acetonitrile in ammonium acetate buffer (10 mM, pH 4.5)] at a flow rate of 0.7 mL/min. Mass spectrometric detection was performed in positive electrospray ionization mode using multiple reaction monitoring (RBT, m/z 847.7 → 815.4; LPV, m/z 629.6 → 447.4). Raloxifene and phenacetin were used as internal standards for RBT and LPV, respectively. Linearity was established in the range of 1-1,000 ng/mL and 0.5-10 µg/mL (R2 ≥ 0.99) for RBT and LPV, respectively. The recovery of LPV and RBT were always >90 and >50%, respectively. The precisions and accuracies were well within the acceptable limits of variation.
Product Description
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 Prep Guard Cartridge, 10 μm particle size, L × I.D. 1 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 10 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm