Merck
  • Mutant IDH1 Downregulates ATM and Alters DNA Repair and Sensitivity to DNA Damage Independent of TET2.
Mutant IDH1 Downregulates ATM and Alters DNA Repair and Sensitivity to DNA Damage Independent of TET2.
Cancer cell (2016-07-19)
Satoshi Inoue, Wanda Y Li, Alan Tseng, Isabel Beerman, Andrew J Elia, Sean C Bendall, François Lemonnier, Ken J Kron, David W Cescon, Zhenyue Hao, Evan F Lind, Naoya Takayama, Aline C Planello, Shu Yi Shen, Alan H Shih, Dana M Larsen, Qinxi Li, Bryan E Snow, Andrew Wakeham, Jillian Haight, Chiara Gorrini, Christian Bassi, Kelsie L Thu, Kiichi Murakami, Alisha R Elford, Takeshi Ueda, Kimberly Straley, Katharine E Yen, Gerry Melino, Luisa Cimmino, Iannis Aifantis, Ross L Levine, Daniel D De Carvalho, Mathieu Lupien, Derrick J Rossi, Garry P Nolan, Rob A Cairns, Tak W Mak
ABSTRACT
Mutations in the isocitrate dehydrogenase-1 gene (IDH1) are common drivers of acute myeloid leukemia (AML) but their mechanism is not fully understood. It is thought that IDH1 mutants act by inhibiting TET2 to alter DNA methylation, but there are significant unexplained clinical differences between IDH1- and TET2-mutant diseases. We have discovered that mice expressing endogenous mutant IDH1 have reduced numbers of hematopoietic stem cells (HSCs), in contrast to Tet2 knockout (TET2-KO) mice. Mutant IDH1 downregulates the DNA damage (DD) sensor ATM by altering histone methylation, leading to impaired DNA repair, increased sensitivity to DD, and reduced HSC self-renewal, independent of TET2. ATM expression is also decreased in human IDH1-mutated AML. These findings may have implications for treatment of IDH-mutant leukemia.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse