Merck
  • Multiplexed Thiol Reactivity Profiling for Target Discovery of Electrophilic Natural Products.
Multiplexed Thiol Reactivity Profiling for Target Discovery of Electrophilic Natural Products.
Cell chemical biology (2017-10-11)
Caiping Tian, Rui Sun, Keke Liu, Ling Fu, Xiaoyu Liu, Wanqi Zhou, Yong Yang, Jing Yang
ABSTRACT
Electrophilic groups, such as Michael acceptors, expoxides, are common motifs in natural products (NPs). Electrophilic NPs can act through covalent modification of cysteinyl thiols on functional proteins, and exhibit potent cytotoxicity and anti-inflammatory/cancer activities. Here we describe a new chemoproteomic strategy, termed multiplexed thiol reactivity profiling (MTRP), and its use in target discovery of electrophilic NPs. We demonstrate the utility of MTRP by identifying cellular targets of gambogic acid, an electrophilic NP that is currently under evaluation in clinical trials as anticancer agent. Moreover, MTRP enables simultaneous comparison of seven structurally diversified α,β-unsaturated γ-lactones, which provides insights into the relative proteomic reactivity and target preference of diverse structural scaffolds coupled to a common electrophilic motif and reveals various potential druggable targets with liganded cysteines. We anticipate that this new method for thiol reactivity profiling in a multiplexed manner will find broad application in redox biology and drug discovery.
Sigma-Aldrich
iTRAQ® Reagent - 8PLEX Multiplex Kit
Sigma-Aldrich
Anti-Importin α antibody, Mouse monoclonal, clone IM-75, purified from hybridoma cell culture
Sigma-Aldrich
MDH2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%