Merck
  • Automatic single-step quick, easy, cheap, effective, rugged and safe sample preparation devices for analysis of pesticide residues in foods.
Automatic single-step quick, easy, cheap, effective, rugged and safe sample preparation devices for analysis of pesticide residues in foods.
Journal of chromatography. A (2017-09-25)
Jishi Wang, Zeying He, Lu Wang, Yaping Xu, Yi Peng, Xiaowei Liu
ABSTRACT
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and improve sample preparation efficiency. This combination integrates QuEChERS procedures into a single run via the use of a vortex vibration-centrifuge device and a centrifuge filtration tube. To validate the efficiency of our automatic QuEChERS device, 270 pesticides were analyzed in plant origined foods including celery, tomatoes, leeks, eggplants, grapes, corn, green tea, and soybean oil using this automatic platform. The results were then compared with those obtained using the manual QuEChERS method. Different parameters were validated and compared including recovery, linearity, repeatability and limits of quantification (LOQ). Satisfactory results, comparable to results obtained using the manual QuEChERS method were obtained. The average recoveries ranged between 70% and 120% for most pesticides with associated relative standard deviations (RSDs) <20% (n=5) indicating satisfactory accuracy and repeatability. An LOQ of 2μg/kg was obtained for most pesticides present in celery and corn matrices, and the correlation coefficients (r2) were >0.990 within a linearity range of 2-500μg/kg. Compared to manual QuEChERS, this novel automatic QuEChERS device and combination could significantly improve the sample preparation efficiency for the multiresidue analysis of pesticides.
Product Description
Supelco
Dimepiperate, PESTANAL®, analytical standard
Supelco
Disulfoton-sulfoxide, PESTANAL®, analytical standard
Supelco
Dipropetryn, PESTANAL®, analytical standard
Supelco
Prothiofos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Disulfoton-sulfone, PESTANAL®, analytical standard
Supelco
Cyprodinil, PESTANAL®, analytical standard
Supelco
Anilofos, PESTANAL®, analytical standard
Supelco
Fenthion-sulfone, PESTANAL®, analytical standard