Merck
  • Acetylcholine-related proteins in non-neoplastic appearing colonic mucosa from patients with colorectal neoplasia.
Acetylcholine-related proteins in non-neoplastic appearing colonic mucosa from patients with colorectal neoplasia.
Molecular carcinogenesis (2017-05-26)
Morten Matthiesen Bach Damm, Thorbjørn Søren Rønn Jensen, Badar Mahmood, Morten Lundh, Steen Seier Poulsen, Niels Bindslev, Mark Berner Hansen
ABSTRACT
The pathogenesis of colorectal neoplasia (CRN) has been associated with altered non-neuronal acetylcholine (ACh) metabolism. The aim of this study was to characterize expression, function, and cellular location of ACh-related proteins in biopsies obtained from endoscopic normal-appearing sigmoid colon in patients with and without CRN. Messenger-RNA (mRNA) levels of 17 ACh-related proteins were quantified by rt-qPCR. Functional responses to ACh, measured as electrogenic transepithelial short circuit current (SCC), were recorded using the Ussing chamber technique. Finally, cellular localization of choline transporter-like proteins (CTLs) and butyryl-cholinesterase enzyme (BChE) was determined by immunohistochemistry. mRNA expression of CTL1 and CTL4 was increased in patients with CRN (P = 0.002 and P = 0.04, respectively). In functional experiments, baseline SCC was increased in CRN patients. ACh induced rapid biphasic changes in SCC. An initial decreasing phase was observed in the minority of CRN patients versus the majority of controls (25% vs 69%, respectively, P = 0.031). For the second increasing phase of SCC, data indicated ACh-activation of two receptors. For both parts of the biphasic response, the half maximal effective concentration and maximal responses showed no difference between patient groups. Immunohistochemistry demonstrated CTL1, 3 and 4 and BChE to be localized to colonic crypt cells. We conclude that CRN is associated with increased expression of CTL1 and CTL4, augmented basal prostaglandin-dependent secretion, and altered functional channel response to ACh in human endoscopic normal-appearing colonic mucosa. The immunohistochemical findings support CTL1, CTL3, CTL4, and BChE to be involved in non-neuronal mucosal ACh metabolism.
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-BCHE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SLC44A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Anti-SLC44A4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Ouabain, European Pharmacopoeia (EP) Reference Standard