Merck
Specificity and mechanism of mandelamide hydrolase catalysis.
Archives of biochemistry and biophysics (2017-01-29)
S A Adediran, Pan-Fen Wang, Abbas G Shilabin, Charles A Baron, Michael J McLeish, R F Pratt
ABSTRACT
The best-studied amidase signature (AS) enzyme is probably fatty acid amide hydrolase (FAAH). Closely related to FAAH is mandelamide hydrolase (MAH), whose substrate specificity and mechanism of catalysis are described in this paper. First, we developed a convenient chromogenic substrate, 4-nitrophenylacetamide, for MAH. The lack of reactivity of MAH with the corresponding ethyl ester confirmed the very limited size of the MAH leaving group site. The reactivity of MAH with 4-nitrophenyl acetate and methyl 4-nitrophenyl carbonate, therefore, suggested formation of an "inverse" acyl-enzyme where the small acyl-group occupies the normal leaving group site. We have interpreted the specificity of MAH for phenylacetamide substrates and small leaving groups in terms of its active site structure, using a homology model based on a FAAH crystal structure. The relevant structural elements were compared with those of FAAH. Phenylmethylboronic acid is a potent inhibitor of MAH (K
Sigma-Aldrich
Phenyl carbamate, 97%
Sigma-Aldrich
4-Nitroacetanilide, 98%
Sigma-Aldrich
(Trimethylsilyl)isocyanate, 85%