Merck
  • USP9X regulates centrosome duplication and promotes breast carcinogenesis.
USP9X regulates centrosome duplication and promotes breast carcinogenesis.
Nature communications (2017-04-01)
Xin Li, Nan Song, Ling Liu, Xinhua Liu, Xiang Ding, Xin Song, Shangda Yang, Lin Shan, Xing Zhou, Dongxue Su, Yue Wang, Qi Zhang, Cheng Cao, Shuai Ma, Na Yu, Fuquan Yang, Yan Wang, Zhi Yao, Yongfeng Shang, Lei Shi
ABSTRACT
Defective centrosome duplication is implicated in microcephaly and primordial dwarfism as well as various ciliopathies and cancers. Yet, how the centrosome biogenesis is regulated remains poorly understood. Here we report that the X-linked deubiquitinase USP9X is physically associated with centriolar satellite protein CEP131, thereby stabilizing CEP131 through its deubiquitinase activity. We demonstrate that USP9X is an integral component of centrosome and is required for centrosome biogenesis. Loss-of-function of USP9X impairs centrosome duplication and gain-of-function of USP9X promotes centrosome amplification and chromosome instability. Significantly, USP9X is overexpressed in breast carcinomas, and its level of expression is correlated with that of CEP131 and higher histologic grades of breast cancer. Indeed, USP9X, through regulation of CEP131 abundance, promotes breast carcinogenesis. Our experiments identify USP9X as an important regulator of centrosome biogenesis and uncover a critical role for USP9X/CEP131 in breast carcinogenesis, supporting the pursuit of USP9X/CEP131 as potential targets for breast cancer intervention.
Sigma-Aldrich
Monoclonal Anti-USP9X antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human PCM1
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
c-Myc Peptide, lyophilized powder
Sigma-Aldrich
Anti-ITCH antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human USP9X
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone CMA312, clone CMA312, from mouse
Sigma-Aldrich
MG-132(R), ≥95% (HPLC)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution