Merck
  • Small polyanion recognition of a triazolium cyclodextrin click cluster in water.
Small polyanion recognition of a triazolium cyclodextrin click cluster in water.
Organic & biomolecular chemistry (2015-07-04)
Hoa Thi Le, Seung Cheol Park, Chulhun Kang, Choon Woo Lim, Tae Woo Kim
ABSTRACT
In order to detect small polyanions (sPAs), which play important roles in many biological systems, a triazolium cyclodextrin click cluster (5, hexakis{6-(3-methyl-4-hydroxymethyl-1H-1,2,3-triazolium-1-yl)-6-deoxy}-α-cyclodextrin iodide) was synthesized and characterized. The competition binding to 5 occupied by 5-carboxyfluorescein of inositol-1,4,5-trisphosphate (IP3), phytic acid, adenosine triphosphate (ATP), ethylenediaminetetraacetic acid (EDTA), glucose, and glucose-6-phosphate was evaluated by UV/vis titration in HEPES (10 mM, pH 7.4) : methanol (1 : 1, v/v). We obtained the binding constants of IP3 and phytic acid to 5 (1.4 × 10(6) and 1.9 × 10(6) M(-1), respectively); however, the binding constants of ATP and EDTA were significantly lower (2.1 × 10(5) and 4.5 × 10(4) M(-1), respectively). Moreover, glucose and glucose-6-phosphate did not show any detectable binding. In addition, the sPA recognition of the triazolium cyclodextrin click cluster in water was confirmed by fluorescence titration.
Sigma-Aldrich
5-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Iodomethane, SAJ special grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
D-myo-Inositol 1,4,5-tris-phosphate trisodium salt, ≥95% (by 1H NMR and TLC)