Merck
  • Functional Characterization of the Receiver Domain for Phosphorelay Control in Hybrid Sensor Kinases.
Functional Characterization of the Receiver Domain for Phosphorelay Control in Hybrid Sensor Kinases.
PloS one (2015-07-08)
Emiko Kinoshita-Kikuta, Eiji Kinoshita, Yoko Eguchi, Shiho Yanagihara, Keisuke Edahiro, Yuki Inoue, Momoka Taniguchi, Myu Yoshida, Kaneyoshi Yamamoto, Hirotaka Takahashi, Tatsuya Sawasaki, Ryutaro Utsumi, Tohru Koike
ABSTRACT
Hybrid sensor kinase, which contains a histidine kinase (HK) domain, a receiver domain, and a histidine-containing phosphotransmitter (HPt) domain, conveys signals to its cognate response regulator by means of a His-Asp-His-Asp phosphorelay. We examined the multistep phosphorelay of a recombinant EvgAS system in Escherichia coli and performed in vitro quantitative analyses of phosphorylation by using Phos-tag SDS-PAGE. Replacement of Asp in the receiver domain of EvgS by Ala markedly promoted phosphorylation at His in the HK domain compared with that in wild-type EvgS. Similar Ala-substituted mutants of other hybrid sensor kinases BarA and ArcB showed similar characteristics. In the presence of sufficient ATP, autophosphorylation of the HK domain in the mutant progressed efficiently with nearly pseudo-first-order kinetics until the phosphorylation ratio reached a plateau value of more than 95% within 60 min, and the value was maintained until 180 min. However, both wild-type EvgS and the Ala-substituted mutant of His in the HPt domain showed a phosphorylation ratio of less than 25%, which gradually decreased after 10 min. These results showed that the phosphorylation level is regulated negatively by the receiver domain. Furthermore, our in vivo assays confirmed the existence of a similar hyperphosphorylation reaction in the HK domain of the EvgS mutant in which the Asp residue was replaced with Ala, confirming the validity of the control mechanism proposed from profiling of phosphorylation in vitro [corrected].
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
L-(+)-Arabinose, 99%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O