Merck
  • Transnitrosation of non-mutagenic N-nitrosoproline forms mutagenic N-nitroso-N-methylurea.
Transnitrosation of non-mutagenic N-nitrosoproline forms mutagenic N-nitroso-N-methylurea.
Bioorganic & medicinal chemistry (2015-05-16)
Keiko Inami, Junko Shiino, Shin Hagiwara, Kei Takeda, Masataka Mochizuki
ABSTRACT
N-Nitroso-N-methylurea (NMU) is a potent carcinogen and suspected as a cause of human cancer. In this study, mutagenic NMU was detected by HPLC after the transnitrosation of non-mutagenic N-nitrosoproline (NP) to N-methylurea in the presence of thiourea (TU) under acidic conditions. The structure of NMU was confirmed by comparing (1)H NMR and IR spectra with that of authentic NMU after fractionation by column chromatography. Furthermore, a fraction containing NMU formed by transnitrosation was mutagenic in Salmonella typhimurium TA1535. NMU was formed in the reaction of NP and N-methylurea in the presence of 1,1,3,3-tetramethylthiourea (TTU) or 1,3-dimethylthiourea in place of TU as an accelerator. The reaction rate constants (k) for NMU formation were correlated with their nucleophilicity of sulfur atom in thioureas. The N-methylurea concentration did not affect the NMU formation, whereas the rate of NMU formation correlated linearly with concentrations of NP, TTU and oxonium ion. The observed kinetics suggests a mechanism by which the nitroso group was transferred directly from the protonated NP to the thiourea then to N-methylurea to form NMU. The rate-determining step was the formation of the complex with the protonated NP and thiourea.
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Ammonium sodium phosphate dibasic tetrahydrate, BioXtra, ≥99%
Sigma-Aldrich
Ammonium sodium phosphate dibasic tetrahydrate, ≥99%
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Proline
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, SAJ special grade, ≥99.0%