Merck
  • A drug formulation using an alginate hydrogel matrix for efficient oral delivery of the manganese porphyrin-based superoxide dismutase mimic.
A drug formulation using an alginate hydrogel matrix for efficient oral delivery of the manganese porphyrin-based superoxide dismutase mimic.
Biomaterials science (2015-07-30)
T Aikawa, S Ito, M Shinohara, M Kaneko, T Kondo, M Yuasa
ABSTRACT
In order for patients to avail of the therapeutic benefits of antioxidant drugs efficiently and conveniently, a robust oral delivery system needs to be developed. However, a common problem in oral drug delivery is ensuring that the drug remains functionally intact even after it has passed through the acidic environment of the gastrointestinal (GI) tract. To protect drugs within the GI environment, we formulated a design based on encapsulating liposomal drugs by using an alginate matrix as a carrier. The liposomal drug was composed of manganese porphyrin (Mn-por), which has been developed as a mimic of superoxide dismutase (SOD), as the therapeutic agent based on the antioxidative effect, namely superoxide (O2˙(-)) inhibitory activity. A cytochrome c assay revealed that the O2˙(-) inhibitory activity of Mn-por could be maintained even after treatment with simulated gastric and intestinal fluids. We demonstrated that oral administration of the formulated drug significantly inhibited the growth of transplanted tumors in mice. The drug formulation presented in this study would be a good candidate for orally available systems, which can effectively deliver SOD mimics.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sodium stearate, ≥99%
SAFC
Cholesterol, Plant-Derived, SyntheChol®