Merck
  • Adding Amorphous Content to Highly Crystalline Polymer Nanowire Solar Cells Increases Performance.
Adding Amorphous Content to Highly Crystalline Polymer Nanowire Solar Cells Increases Performance.
Advanced materials (Deerfield Beach, Fla.) (2015-05-06)
Han Yan, Yin Song, George R McKeown, Gregory D Scholes, Dwight S Seferos
ABSTRACT
Polymer solar cells are fabricated with systematic variation of the phase purity. Photovoltaic tests demonstrate that devices with ca. 10% of mixed phases outperform pure-phase devices. Photophysical studies reveal the effects of mixed phase on charge generation and recombination. These results show a promising strategy for the optimization of organic electronic materials.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free