Merck
  • Effects of phytochemicals on in vitro anti-inflammatory activity of Bifidobacterium adolescentis.
Effects of phytochemicals on in vitro anti-inflammatory activity of Bifidobacterium adolescentis.
Bioscience, biotechnology, and biochemistry (2015-02-28)
Kyuichi Kawabata, Yuri Kato, Taiken Sakano, Nobuyuki Baba, Kota Hagiwara, Akira Tamura, Seigo Baba, Midori Natsume, Hajime Ohigashi
ABSTRACT
Probiotics have been shown to improve the condition of not only the human gastrointestinal tract but also the entire body. We found that quercetin enhances the anti-inflammatory activity of Bifidobacterium adolescentis, which is abundant in human intestines. Here, we assessed whether certain phytochemicals could enhance the anti-inflammatory activity of B. adolescentis. Bifidobacteria were anaerobically cultured with phytochemicals for 3 h, and the anti-inflammatory activity of the supernatants was estimated by testing their ability to inhibit nitric oxide (NO) production by lipopolysaccharide-stimulated RAW264 macrophages. Of the 55 phytochemicals tested, phloretin, (+)-taxifolin, and (-)-epigallocatechin gallate as well as quercetin-3-O-glucoside and quercetin-4'-O-glucoside were similar to quercetin in promoting NO suppression by B. adolescentis. In addition, the phytochemicals excluding quercetin increased the concentrations of lactic and acetic acids in the co-culture supernatants. These results suggest that some phytochemicals may activate the anti-inflammatory function of B. adolescentis.
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Flavone, ≥99.0%
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
Resorcinol, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Resorcinol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
3′,4′-Dihydroxyflavonol, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea