Merck
  • Sensitivity enhancement of pH indicator and its application in the evaluation of fish freshness.
Sensitivity enhancement of pH indicator and its application in the evaluation of fish freshness.
Talanta (2015-06-17)
Wen Sun, Hui Li, Han Wang, Shan Xiao, Jihui Wang, Liang Feng
ABSTRACT
The sensitivity of pH indicators is always confined in acidic solvents used during the immobilization because of the large deviation from their color transition points. By regulating the pH of indicators using organic acid or base, the color transition points of indicators could be readily achieved and the sensitivity could be greatly enhanced. In this study, bromophenol blue was selected as an example to interpret this improvement. And Hue, Saturation and Value (HSV) color model was applied to obtain digital color information of pH indicators. In addition, other two kinds of pH indicators were chosen and regulated to their color transition points as well. The resultant indicators together with the original ones were composed to a colorimetric sensor array and utilized to determine fish freshness. Compared with the original indicators, much higher sensitivity in the detection of fish freshness was obtained by the pH regulated indicators.
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
Triethoxy(octyl)silane, deposition grade, 97%, 99.99% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
2-Methoxyethanol, JIS special grade, ≥99.0%
Sigma-Aldrich
2-Methoxyethanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Triethoxy(octyl)silane, 97%
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems