Merck
  • Dual Stimuli-Responsive Poly(β-amino ester) Nanoparticles for On-Demand Burst Release.
Dual Stimuli-Responsive Poly(β-amino ester) Nanoparticles for On-Demand Burst Release.
Macromolecular bioscience (2015-06-04)
Jung Seok Lee, Xiaojian Deng, Patrick Han, Jianjun Cheng
ABSTRACT
We designed poly(β-amino esters) (PBAEs) bearing both UV light- and pH-sensitive groups and used PBAEs to prepare nanoparticles (NPs) that can be utilized for on-demand burst release of guest molecules in response to multiple triggers. Due to the presence of the photo-cleavable group in each repeating unit of PBAE, rapid release of encapsulated model drug could be achieved even with exposures to low intensity UV (10 mW · cm(-2) ). Especially, the burst release was further accelerated by additional UV treatments in the acidic condition showing the combinatory effect of dual stimuli. We believe these PBAE-based NPs can potentially be used to design intelligent controlled release device and nanomedicines.
Sigma-Aldrich
Potassium permanganate solution, 0.1 M
Sigma-Aldrich
Potassium permanganate solution, 0.01 M
Sigma-Aldrich
Potassium permanganate solution, 0.2 M
Sigma-Aldrich
Potassium permanganate solution, 0.02 M
Sigma-Aldrich
Potassium permanganate solution, 5 mM
Sigma-Aldrich
Potassium permanganate solution, 2 mM
Sigma-Aldrich
Borane tetrahydrofuran complex solution, 1.0 M in THF, contains 0.005 M N-Isopropyl-N-methyl-tert-butylamine as stabilizer
Sigma-Aldrich
1,3-Dimethyl-2-nitrobenzene, 99%
Sigma-Aldrich
Borane tetrahydrofuran complex solution, 1.0 M in THF
Sigma-Aldrich
Acryloyl chloride, 97.0%, contains <210 ppm MEHQ as stabilizer
Sigma-Aldrich
Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer
Sigma-Aldrich
4-Amino-1-butanol, 98%