Merck
  • Sorting of Clathrin-Independent Cargo Proteins Depends on Rab35 Delivered by Clathrin-Mediated Endocytosis.
Sorting of Clathrin-Independent Cargo Proteins Depends on Rab35 Delivered by Clathrin-Mediated Endocytosis.
Traffic (Copenhagen, Denmark) (2015-05-20)
Dipannita Dutta, Julie G Donaldson
ABSTRACT
Clathrin-mediated endocytosis (CME) and clathrin-independent endocytosis (CIE) co-exist in most cells but little is known about their communication and coordination. Here we show that when CME was inhibited, endocytosis by CIE continued but endosomal trafficking of CIE cargo proteins was altered. CIE cargo proteins that normally traffic directly into Arf6-associated tubules after internalization and avoid degradation (CD44, CD98 and CD147) now trafficked to lysosomes and were degraded. The endosomal tubules were also absent and Arf6-GTP levels were elevated. The altered trafficking, loss of the tubular endosomal network and elevated Arf6-GTP levels caused by inhibition of CME were rescued by expression of Rab35, a Rab associated with clathrin-coated vesicles, or its effector ACAPs, Arf6 GTPase activating proteins (GAP) that inactivate Arf6. Furthermore, siRNA knockdown of Rab35 recreated the phenotype of CME ablation on CIE cargo trafficking without altering endocytosis of transferrin. These observations suggest that Rab35 serves as a CME detector and that loss of CME, or Rab35 input, leads to elevated Arf6-GTP and shifts the sorting of CIE cargo proteins to lysosomes and degradation.
Sigma-Aldrich
Xenon-132Xe, 99.5 atom %
Sigma-Aldrich
MISSION® esiRNA, targeting human CLTC
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cltc
Sigma-Aldrich
Xenon-132Xe, 60 atom %
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB35
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rab35