Merck
  • Pseudomonas spp. and Serratia liquefaciens as Predominant Spoilers in Cold Raw Milk.
Pseudomonas spp. and Serratia liquefaciens as Predominant Spoilers in Cold Raw Milk.
Journal of food science (2015-07-21)
Solimar G Machado, Fernanda L da Silva, Denise M S Bazzolli, Marc Heyndrickx, Paulo M de A Costa, Maria Cristina D Vanetti
ABSTRACT
The storage of fresh raw milk at low temperature does not prevent proliferation of psychrotrophic bacteria that can produce heat-resistant proteolytic enzymes contributing to the reduced shelf life of dairy products. This study aimed to identify the dominant psychrotrophic proteolytic enzyme-producing population of raw milk from Brazil. Raw milk samples collected in 3 different cooling tanks in Brazil were stored at optimal (45 h at 4 °C followed by 3 h at 7 °C) and suboptimal (45 h at 7 °C followed by 3 h at 10 °C) conditions to simulate farm storage and transportation allowed by Brazilian laws. The highly proteolytic enzyme-producing strains isolated from stored cold raw milk were characterized by repetitive sequence-based Polymerase Chain Reaction (PCR) analysis. This clustering resulted in 8 different clusters and 4 solitary fingerprints. The most proteolytic isolates from each rep-cluster were selected for identification using miniaturized kit, 16S rDNA and rpoB gene sequencing. Serratia liquefaciens (73.9%) and Pseudomonas spp. (26.1%) were identified as the dominant psychrotrophic microorganisms with high spoilage potential. The knowledge of milk spoilage microbiota will contribute to improved quality of milk and dairy products.
Sigma-Aldrich
Bromothymol Blue solution, pH 6.0-7.6
Sigma-Aldrich
Bromothymol Blue solution, pH 6.0-7.6
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Bromothymol Blue, ACS reagent, Dye content 95 %
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Bromothymol Blue, JIS special grade
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Fusidic acid sodium salt, ≥98% (TLC)
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Potassium sulfate, suitable for plant cell culture
Sigma-Aldrich
Potassium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium sulfate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
SAFC
Potassium sulfate