Merck
  • Development of full-thickness human skin equivalents with blood and lymph-like capillary networks by cell coating technology.
Development of full-thickness human skin equivalents with blood and lymph-like capillary networks by cell coating technology.
Journal of biomedical materials research. Part A (2015-04-09)
Michiya Matsusaki, Kumiko Fujimoto, Yuji Shirakata, Satoshi Hirakawa, Koji Hashimoto, Mitsuru Akashi
ABSTRACT
We developed a human skin equivalent (HSE) containing blood and lymph-like capillary networks using a cell coating technique, which is a rapid fabrication technology of three-dimensional cellular constructs by cell surface coating using layer-by-layer assembled nanofilms of extracellular matrices. The thickness of dermis consisting of normal human dermal fibroblasts was easily controlled from approximately 5 to 100 µm by altering the seeded cell number. Keratinocytes as a major cell population showed homogeneous differentiation on the surface of the dermis by lifting to air-liquid interface. Histological analysis revealed four distinct layers such as basal layer, spinous layer, granular layer, and cornified cell layer in the epidermis. Interestingly, the measurement of transepithelial electrical resistance (TEER) indicated prolongation of the attainment time for maximum value by increasing the number of the dermal fibroblasts, and the HSEs with six layers of dermis revealed the longest period maintaining over 500 Ω cm(2) of TEER. The co-sandwich culture of human umbilical vein endothelial cells and normal human dermal lymphatic microvascular endothelial cells within eight-layered dermis showed in vitro co-network formation of individual blood and lymph-like capillaries inside the dermis. This is the report for homogeneous full-thickness HSEs with blood and lymph capillary networks, which will be useful for biomedical and pharmaceutical applications.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%