Merck
  • Synthesis of a novel 4H-pyran analog as minor groove binder to DNA using ethidium bromide as fluorescence probe.
Synthesis of a novel 4H-pyran analog as minor groove binder to DNA using ethidium bromide as fluorescence probe.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-07-25)
M M V Ramana, Rahul Betkar, Amey Nimkar, Prasanna Ranade, Balaji Mundhe, Sachin Pardeshi
ABSTRACT
In the present work, isopropyl-6-amino-4-(3,5-bis(trifluoromethyl)phenyl)-5-cyano-2-methyl-4H-pyran-3-carboxylate (4H-pyran analog) has been synthesized by a three component reaction catalyzed by CsOH/γ-Al2O3 and characterized. The interaction of 4H-pyran analog with herring sperm DNA (hs DNA) under physiological conditions (phosphate buffer of pH 7.2) was investigated by UV absorption, FT-IR, fluorescence, (31)P NMR and circular dichroism (CD) spectroscopy. Fluorescence quenching results reveal that static quenching mechanism is involved in binding between 4H-pyran analog and hs DNA. The calculated thermodynamic parameters (ΔH° and ΔS°) indicate that hydrogen bonding plays a major role in binding between them. UV absorption and fluorescence shows the binding mode of 4H-pyran analog with hs DNA as non-intercalative. According to the IR spectroscopy, 4H-pyran analog binds to guanine, thymine, adenine bases of hs DNA but not to phosphate backbone of hs DNA which is also in good agreement with (31)P NMR results. CD and competitive binding experiment results confirms the minor groove binding of 4H-pyran analog to hs DNA.
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
Isopropyl acetoacetate, Arxada quality, ≥99.0% (GC)
Sigma-Aldrich
Malononitrile, Arxada quality, ≥99.0% (calculated, GC, KF)
Sigma-Aldrich
Malononitrile, ≥99%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Cesium hydroxide solution, 50 wt. % in H2O, 99% trace metals basis
Sigma-Aldrich
Cesium hydroxide solution, 50 wt. % in H2O, 99.9% trace metals basis
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
Supelco
Pyridaben, PESTANAL®, analytical standard