Merck
  • Determination of Fatty Acid Metabolism with Dynamic [11C]Palmitate Positron Emission Tomography of Mouse Heart In Vivo.
Determination of Fatty Acid Metabolism with Dynamic [11C]Palmitate Positron Emission Tomography of Mouse Heart In Vivo.
Molecular imaging (2015-10-16)
Yinlin Li, Tao Huang, Xinyue Zhang, Min Zhong, Natalie N Walker, Jiang He, Stuart S Berr, Susanna R Keller, Bijoy K Kundu
ABSTRACT
The goal of this study was to establish a quantitative method for measuring fatty acid (FA) metabolism with partial volume (PV) and spill-over (SP) corrections using dynamic [(11)C]palmitate positron emission tomographic (PET) images of mouse heart in vivo. Twenty-minute dynamic [(11)C]palmitate PET scans of four 18- to 20-week-old male C57BL/6 mice under isoflurane anesthesia were performed using a Focus F-120 PET scanner. A model-corrected blood input function, by which the input function with SP and PV corrections and the metabolic rate constants (k1-k5) are simultaneously estimated from the dynamic [(11)C]palmitate PET images of mouse hearts in a four-compartment tracer kinetic model, was used to determine rates of myocardial fatty acid oxidation (MFAO), myocardial FA esterification, myocardial FA use, and myocardial FA uptake. The MFAO thus measured in C57BL/6 mice was 375.03 ± 43.83 nmol/min/g. This compares well to the MFAO measured in perfused working C57BL/6 mouse hearts ex vivo of about 350 nmol/g/min and 400 nmol/min/g. FA metabolism was measured for the first time in mouse heart in vivo using dynamic [(11)C]palmitate PET in a four-compartment tracer kinetic model. MFAO obtained with this model was validated by results previously obtained with mouse hearts ex vivo.
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethyl ether, ≥99.5%
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1