Merck
  • Branched/linear selectivity in palladium-catalyzed allyl-allyl cross-couplings: The role of ligands.
Branched/linear selectivity in palladium-catalyzed allyl-allyl cross-couplings: The role of ligands.
Tetrahedron (2015-08-19)
Michael J Ardolino, James P Morken
ABSTRACT
While Pd-catalyzed allyl-allyl cross-couplings in the presence of small-bite-angle bidentate ligands reliably furnish the branched regioisomer with high levels of selectivity, cross-couplings in the presence of large-bite-angle bidentate ligands give varying, often unpredictable, levels of selectivity. In a combined computational and experimental study, we probe the underlying features that govern the regioselectivity in these metal-catalyzed cross-couplings.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
[Pd2(dba)3] x dba, Umicore
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Xantphos, 97%
Sigma-Aldrich
Tris(dibenzylideneacetone)dipalladium(0), 97%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Tris(dibenzylideneacetone)dipalladium(0) ChemBeads