Merck
Susceptibility of glutinous rice starch to digestive enzymes.
Carbohydrate polymers (2015-05-26)
Li Guo, Juanjuan Zhang, Jian Hu, Xueling Li, Xianfeng Du
ABSTRACT
To understand the susceptibility of glutinous rice starch to digestive enzymes and its potential impact on glycemic response, enzyme kinetics and in vitro digestibility of the native and gelatinized starches were investigated. The results showed that the Km values of the native and gelatinized starch were 10.35 mg/mL and 9.92 mg/mL, respectively. The digestion rate coefficients k values of the native and gelatinized starches were 2.0 × 10(-3)min(-1) and 1.1 × 10(-2)min(-1), respectively. The contents of rapid digestible starch (RDS), slowly digestible starch (SDS) and resistant starch (RS) in native glutinous rice starch were 8.92%, 21.52% and 69.56%, respectively. After gelatinization, the amounts of RDS, SDS and RS were 18.47%, 29.75% and 51.78%, respectively. The native and gelatinized glutinous rice starches were 10.34% and 14.07% for hydrolysis index (HI), as well as 43.14% and 45.92% for glycemic index (GI), respectively. During the in vitro digestion, the crystallinity of native glutinous rice starch was increased from 34.7% to 35.8% and 38.4% after 20 and 120 min, respectively.
Sigma-Aldrich
D-(+)-Maltose monohydrate, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(+)-Maltose monohydrate, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(+)-Maltose monohydrate, ≥98% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
D-(+)-Maltose monohydrate, from potato, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Maltose monohydrate, Type II, ≥95%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
D-(+)-Maltose monohydrate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M