Merck
  • PRMT1-mediated methylation of the EGF receptor regulates signaling and cetuximab response.
PRMT1-mediated methylation of the EGF receptor regulates signaling and cetuximab response.
The Journal of clinical investigation (2015-11-17)
Hsin-Wei Liao, Jung-Mao Hsu, Weiya Xia, Hung-Ling Wang, Ying-Nai Wang, Wei-Chao Chang, Stefan T Arold, Chao-Kai Chou, Pei-Hsiang Tsou, Hirohito Yamaguchi, Yueh-Fu Fang, Hong-Jen Lee, Heng-Huan Lee, Shyh-Kuan Tai, Mhu-Hwa Yang, Maria P Morelli, Malabika Sen, John E Ladbury, Chung-Hsuan Chen, Jennifer R Grandis, Scott Kopetz, Mien-Chie Hung
ABSTRACT
Posttranslational modifications to the intracellular domain of the EGFR are known to regulate EGFR functions; however, modifications to the extracellular domain and their effects remain relatively unexplored. Here, we determined that methylation at R198 and R200 of the EGFR extracellular domain by protein arginine methyltransferase 1 (PRMT1) enhances binding to EGF and subsequent receptor dimerization and signaling activation. In a mouse orthotopic colorectal cancer xenograft model, expression of a methylation-defective EGFR reduced tumor growth. Moreover, increased EGFR methylation sustained signaling activation and cell proliferation in the presence of the therapeutic EGFR monoclonal antibody cetuximab. In colorectal cancer patients, EGFR methylation level also correlated with a higher recurrence rate after cetuximab treatment and reduced overall survival. Together, these data indicate that R198/R200 methylation of the EGFR plays an important role in regulating EGFR functionality and resistance to cetuximab treatment.
Sigma-Aldrich
PRMT1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
PRMT1 human, recombinant, expressed in E. coli, ≥50% (SDS-PAGE)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
PRMT1 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit
SAFC
Isopropyl β-D-1-thiogalactopyranoside