Merck
  • STX13 regulates cargo delivery from recycling endosomes during melanosome biogenesis.
STX13 regulates cargo delivery from recycling endosomes during melanosome biogenesis.
Journal of cell science (2015-07-26)
Riddhi Atul Jani, Latha Kallur Purushothaman, Shikha Rani, Ptissam Bergam, Subba Rao Gangi Setty
ABSTRACT
Melanosomes are a class of lysosome-related organelles produced by melanocytes. Biogenesis of melanosomes requires the transport of melanin-synthesizing enzymes from tubular recycling endosomes to maturing melanosomes. The SNARE proteins involved in these transport or fusion steps have been poorly studied. We found that depletion of syntaxin 13 (STX13, also known as STX12), a recycling endosomal Qa-SNARE, inhibits pigment granule maturation in melanocytes by rerouting the melanosomal proteins such as TYR and TYRP1 to lysosomes. Furthermore, live-cell imaging and electron microscopy studies showed that STX13 co-distributed with melanosomal cargo in the tubular-vesicular endosomes that are closely associated with the maturing melanosomes. STX family proteins contain an N-terminal regulatory domain, and deletion of this domain in STX13 increases both the SNARE activity in vivo and melanosome cargo transport and pigmentation, suggesting that STX13 acts as a fusion SNARE in melanosomal trafficking pathways. In addition, STX13-dependent cargo transport requires the melanosomal R-SNARE VAMP7, and its silencing blocks the melanosome maturation, reflecting a defect in endosome-melanosome fusion. Moreover, we show mutual dependency between STX13 and VAMP7 in regulating their localization for efficient cargo delivery to melanosomes.
Sigma-Aldrich
MISSION® esiRNA, targeting human VAMP7 (3)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mecp2
Sigma-Aldrich
3-Amino-1,2,4-triazole, ≥95% (TLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Yeast Synthetic Drop-out Medium Supplements, without leucine and tryptophan
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
4H-1,2,4-Triazol-3-amine, AldrichCPR
Sigma-Aldrich
MISSION® esiRNA, targeting human VAMP7 (4)
Sigma-Aldrich
MISSION® esiRNA, targeting human VAMP7 (1)
Sigma-Aldrich
MISSION® esiRNA, targeting human VAMP7 (2)
Sigma-Aldrich
Yeast Synthetic Drop-out Medium Supplements, without histidine, leucine, and tryptophan
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vamp7
Sigma-Aldrich
MISSION® esiRNA, targeting human PITX2
Sigma-Aldrich
MISSION® esiRNA, targeting human MECP2