Merck
  • Phosphatidylinositol binding of Saccharomyces cerevisiae Pdr16p represents an essential feature of this lipid transfer protein to provide protection against azole antifungals.
Phosphatidylinositol binding of Saccharomyces cerevisiae Pdr16p represents an essential feature of this lipid transfer protein to provide protection against azole antifungals.
Biochimica et biophysica acta (2014-07-30)
Roman Holič, Zuzana Simová, Tim Ashlin, Vladimír Pevala, Katarína Poloncová, Dana Tahotná, Eva Kutejová, Shamshad Cockcroft, Peter Griač
ABSTRACT
Pdr16p is considered a factor of clinical azole resistance in fungal pathogens. The most distinct phenotype of yeast cells lacking Pdr16p is their increased susceptibility to azole and morpholine antifungals. Pdr16p (also known as Sfh3p) of Saccharomyces cerevisiae belongs to the Sec14 family of phosphatidylinositol transfer proteins. It facilitates transfer of phosphatidylinositol (PI) between membrane compartments in in vitro systems. We generated Pdr16p(E235A, K267A) mutant defective in PI binding. This PI binding deficient mutant is not able to fulfill the role of Pdr16p in protection against azole and morpholine antifungals, providing evidence that PI binding is critical for Pdr16 function in modulation of sterol metabolism in response to these two types of antifungal drugs. A novel feature of Pdr16p, and especially of Pdr16p(E235A, K267A) mutant, to bind sterol molecules, is observed.
Product Description
Supelco
Squalene, analytical standard
Sigma-Aldrich
Squalene, ≥98%, liquid
USP
Miconazole, United States Pharmacopeia (USP) Reference Standard
Miconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lanosterol, ≥93%, powder