Merck
  • Determination of homocysteine in cerebrospinal fluid as an indicator for surgery treatment in patients with hydrocefalus.
Determination of homocysteine in cerebrospinal fluid as an indicator for surgery treatment in patients with hydrocefalus.
Physiological research (2014-06-09)
L Sosvorová, J Bešťák, M Bičíková, M Mohapl, M Hill, J Kubátová, R Hampl
ABSTRACT
Increased homocysteine levels in serum are typical features of neurodegenerative brain diseases including hydrocephalus. The most frequent therapeutic approach consists of the insertion of a shunt, connecting the brain ventricles to an alternative drainage site. To decide whether the patient should undergo this, the lumbar drainage test is usually carried out to distinguish patients who can benefit from the shunt insertion. In searching for other potential biochemical markers for shunt indication we determined homocysteine levels in CSF during the lumbar drainage test. Homocysteine in CSF was measured during the 5-day lumbar drainage test in 27 patients with normal-pressure hydrocephalus (NPH) and in 25 patients with excluded hydrocephalus. A novelized gas chromatography method with flame ionization detection (GC-FID) was developed and evaluated. During the first two days of lumbar drainage, the levels of CSF homocysteine in NPH patients were significantly higher compared to the controls, while on the fifth day, the homocysteine levels in patients with hydrocephalus reached the level of controls. Determination of CSF homocysteine in patients with confirmed or suspected hydrocephalus may serve as an independent marker for deciding on their further treatment strategy.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium oxalate monohydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%