Merck
  • Metadherin exon 11 skipping variant enhances metastatic spread of ovarian cancer.
Metadherin exon 11 skipping variant enhances metastatic spread of ovarian cancer.
International journal of cancer (2014-10-28)
Stefan Haug, Dominik Schnerch, Sebastian Halbach, Justin Mastroianni, Verónica I Dumit, Marie Follo, Annette Hasenburg, Martin Köhler, Heide Dierbach, Sebastian Herzog, Amelie Proske, Martin Werner, Joern Dengjel, Tilman Brummer, Silke Laßmann, Ralph Wäsch, Robert Zeiser
ABSTRACT
Metastatic ovarian cancer has a dismal prognosis and current chemotherapeutic approaches have very limited success. Metadherin (MTDH) is expressed in human ovarian cancer tissue and its expression inversely correlates with patients overall survival. Consistent with these studies, we observed MTDH expression in tissue specimens of FIGO Stage III ovarian carcinomas (72/83 cases). However, we also observed this in normal human ovarian epithelial (OE) cells, which raised the question of whether MTDH-variants with functional differences exist. We identified a novel MTDH exon 11 skipping variant (MTDHdel) which was seen at higher levels in ovarian cancer compared to benign OE cells. We analyzed MTDH-binding partner interactions and found that 12 members of the small ribosomal subunit and several mRNA binding proteins bound stronger to MTDHdel than to wildtype MTDH which indicates differential effects on gene translation. Knockdown of MTDH in ovarian cancer cells reduced the amount of distant metastases and improved the survival of ovarian cancer-bearing mice. Selective overexpression of the MTDHdel enhanced murine and human ovarian cancer progression and caused a malignant phenotype in originally benign human OE cells. MTDHdel was detectable in microdissected ovarian cancer cells of some human tissue specimens of ovarian carcinomas. In summary, we have identified a novel MTDH exon 11 skipping variant that shows enhanced binding to small ribosomal subunit members and that caused reduced overall survival of ovarian cancer bearing mice. Based on the findings in the murine system and in human tissues, MTDHdel must be considered a major promalignant factor for ovarian cancer.
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Anti-MTDH antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine