Merck
  • The influence of genetic polymorphism of Cyp2c19 isoenzyme on the pharmacokinetics of clopidogrel and its metabolites in patients with cardiovascular diseases.
The influence of genetic polymorphism of Cyp2c19 isoenzyme on the pharmacokinetics of clopidogrel and its metabolites in patients with cardiovascular diseases.
Journal of clinical pharmacology (2014-05-02)
Marta Karaźniewicz-Łada, Dorota Danielak, Błażej Rubiś, Paweł Burchardt, Grzegorz Oszkinis, Franciszek Główka
ABSTRACT
An extensive investigation on pharmacokinetics of clopidogrel and its metabolites as well as pharmacodynamics of the drug was performed in patients with cardiovascular disease carrying various alleles coding CYP2C19 isoenzyme. The influence of non-genetic factors on the clopidogrel response was also studied. Plasma concentrations of clopidogrel, its carboxylic metabolite, and diastereoisomers of a thiol metabolite (the inactive H3 and the active H4) following an administration of 75 mg of the drug were determined in three groups of patients divided with respect to their CYP2C19 genotype: ultrametabolizers, extensive metabolizers, and intermediate metabolizers. The mean peak plasma concentration of H4 in intermediate metabolizers was 3.1- and 2.8-fold lower than that of ultrametabolizers (P = 0.055) and extensive metabolizers (P = 0.026), respectively. The mean H4 area under the curve (AUC0-24 h ) for intermediate metabolizers were significantly lower than that for ultrametabolizers (P = 0.046). Intermediate metabolizers exhibited a significantly higher platelet aggregation than ultrametabolizers and extensive metabolizers (P = 0.035). A multivariate analysis showed that the effect of CYP2C19*2 allele on an ADP-induced platelet aggregation was better pronounced in the presence of non-genetic risk factors (P = 0.008). We concluded that the CYP2C19*2 genotype is the primary determinant of the antiplatelet response to clopidogrel therapy.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
3′-Methoxyacetophenone, 97%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2-Bromo-3′-methoxyacetophenone, 98%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%