Merck
  • Analysis of sulfonamides, trimethoprim, fluoroquinolones, quinolones, triphenylmethane dyes and methyltestosterone in fish and shrimp using liquid chromatography-mass spectrometry.
Analysis of sulfonamides, trimethoprim, fluoroquinolones, quinolones, triphenylmethane dyes and methyltestosterone in fish and shrimp using liquid chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-10-14)
Joseph M Storey, Susan B Clark, Aaron S Johnson, Wendy C Andersen, Sherri B Turnipseed, Jack J Lohne, Robert J Burger, Patrick R Ayres, Justin R Carr, Mark R Madson
ABSTRACT
A liquid chromatography-tandem mass spectrometry (LC-MS/MS) screening method is described for the detection and identification of 26 veterinary drugs in fish and other aquaculture products. The analytes include: 13 sulfonamides, trimethoprim, 3 fluoroquinolones, 3 quinolones, 3 triphenylmethane dyes, 2 leuco dye metabolites, and 1 hormone. In this method, tissue is mixed with EDTA-McIlvaine buffer, double-extracted with acetonitrile, p-toluenesulfonic (p-TSA) acid and N,N,N',N'-tetramethyl-p-phenylenediamine dihydrochloride (TMPD), and analyzed using LC-MS/MS. Inclusion of p-TSA and TMPD in the extraction procedure was critical for simultaneous analysis of dyes with the other groups of veterinary drugs. The proposed procedure was validated as both a quantitative analysis method and as a semi-quantitative screening method for multiple fish and shrimp matrices. The method was applied to eight types of fish (catfish, eel, pangasius, sablefish, tilapia, swai, salmon, and trout) and shrimp at the appropriate level of concern: 10ng/g for sulfonamides, trimethoprim, and quinolones, 5ng/g for fluoroquinolones, 1ng/g for dyes and their metabolites, and 0.4ng/g for methyltestosterone.
Product Description
Supelco
Sulfadoxin, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sulfadoxine, European Pharmacopoeia (EP) Reference Standard
Sulfamethoxypyridazine, European Pharmacopoeia (EP) Reference Standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Methyltestosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Methyltestosterone, European Pharmacopoeia (EP) Reference Standard
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Supelco
Sulfaethoxypyridazine, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material