Merck
  • An integrated in vitro imaging platform for characterizing filarial parasite behavior within a multicellular microenvironment.
An integrated in vitro imaging platform for characterizing filarial parasite behavior within a multicellular microenvironment.
PLoS neglected tropical diseases (2014-11-21)
Timothy Kassis, Henry M Skelton, Iris M Lu, Andrew R Moorhead, J Brandon Dixon
ABSTRACT
Lymphatic Filariasis, a Neglected Tropical Disease, is caused by thread-like parasitic worms, including B. malayi, which migrate to the human lymphatic system following transmission. The parasites reside in collecting lymphatic vessels and lymph nodes for years, often resulting in lymphedema, elephantiasis or hydrocele. The mechanisms driving worm migration and retention within the lymphatics are currently unknown. We have developed an integrated in vitro imaging platform capable of quantifying B. malayi migration and behavior in a multicellular microenvironment relevant to the initial site of worm injection by incorporating the worm in a Polydimethylsiloxane (PDMS) microchannel in the presence of human dermal lymphatic endothelial cells (LECs) and human dermal fibroblasts (HDFs). The platform utilizes a motorized controllable microscope with CO2 and temperature regulation to allow for worm tracking experiments with high resolution over large length and time scales. Using post-acquisition algorithms, we quantified four parameters: 1) speed, 2) thrashing intensity, 3) percentage of time spent in a given cell region and 4) persistence ratio. We demonstrated the utility of our system by quantifying these parameters for L3 B. malayi in the presence of LECs and HDFs. Speed and thrashing increased in the presence of both cell types and were altered within minutes upon exposure to the anthelmintic drug, tetramisole. The worms displayed no targeted migration towards either cell type for the time course of this study (3 hours). When cells were not present in the chamber, worm thrashing correlated directly with worm speed. However, this correlation was lost in the presence of cells. The described platform provides the ability to further study B. malayi migration and behavior.
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
USP
Hydrocortisone acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Octamethyltrisiloxane, 98%
Hydrocortisone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard