Merck
  • Endogenous endothelin 1 mediates angiotensin II-induced hypertrophy in electrically paced cardiac myocytes through EGFR transactivation, reactive oxygen species and NHE-1.
Endogenous endothelin 1 mediates angiotensin II-induced hypertrophy in electrically paced cardiac myocytes through EGFR transactivation, reactive oxygen species and NHE-1.
Pflugers Archiv : European journal of physiology (2013-12-12)
María V Correa, Mariela B Nolly, Claudia I Caldiz, Gladys E Chiappe de Cingolani, Horacio E Cingolani, Irene L Ennis
ABSTRACT
Emerging evidence supports a key role for endothelin-1 (ET-1) and the transactivation of the epidermal growth factor receptor (EGFR) in angiotensin II (Ang II) action. We aim to determine the potential role played by endogenous ET-1, EGFR transactivation and redox-dependent sodium hydrogen exchanger-1 (NHE-1) activation in the hypertrophic response to Ang II of cardiac myocytes. Electrically paced adult cat cardiomyocytes were placed in culture and stimulated with 1 nmol l(-1) Ang II or 5 nmol l(-1) ET-1. Ang II increased ~45 % cell surface area (CSA) and ~37 % [(3)H]-phenylalanine incorporation, effects that were blocked not only by losartan (Los) but also by BQ123 (AT1 and ETA receptor antagonists, respectively). Moreover, Ang II significantly increased ET-1 messenger RNA (mRNA) expression. ET-1 similarly increased myocyte CSA and protein synthesis, actions prevented by the reactive oxygen species scavenger MPG or the NHE-1 inhibitor cariporide (carip). ET-1 increased the phosphorylation of the redox-sensitive ERK1/2-p90(RSK) kinases, main activators of the NHE-1. This effect was prevented by MPG and the antagonist of EGFR, AG1478. Ang II, ET-1 and EGF increased myocardial superoxide production (187 ± 9 %, 149 ± 8 % and 163.7 ± 6 % of control, respectively) and AG1478 inhibited these effects. Interestingly, Los inhibited only Ang II whilst BQ123 cancelled both Ang II and ET-1 actions, supporting the sequential and unidirectional activation of AT1, ETA and EGFR. Based on the present evidence, we propose that endogenous ET-1 mediates the hypertrophic response to Ang II by a mechanism that involves EGFR transactivation and redox-dependent activation of the ERK1/2-p90(RSK) and NHE-1 in adult cardiomyocytes.
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
N-(2-Mercaptopropionyl)glycine
SAFC
Taurine
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
USP
Levocarnitine, United States Pharmacopeia (USP) Reference Standard
USP
Taurine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%