Merck
  • Synthesis of 45S5 Bioglass® via a straightforward organic, nitrate-free sol-gel process.
Synthesis of 45S5 Bioglass® via a straightforward organic, nitrate-free sol-gel process.
Materials science & engineering. C, Materials for biological applications (2014-05-27)
Ehsan Rezabeigi, Paula M Wood-Adams, Robin A L Drew
ABSTRACT
More than four decades after the discovery of 45S5 Bioglass® as the first bioactive material, this composition is still one of the most promising materials in the tissue engineering field. Sol-gel-derived bioactive glasses generally possess improved properties over other bioactive glasses, because of their highly porous microstructure and unique surface chemistry which accelerate hydroxyapatite formation. In the current study, a new combination of precursors with lactic acid as the hydrolysis catalyst have been employed to design an organic, nitrate-free sol-gel procedure for synthesizing of 45S5 Bioglass®. This straightforward route is able to produce fully amorphous submicron particles of this glass with an appropriately high specific surface area on the order of ten times higher than that of the melt-derived glasses. These characteristics are expected to lead to rapid hydroxyapatite formation and consequently more efficient bone bonding.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Calcium L-lactate pentahydrate, SAJ first grade, ≥97.0%
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Calcium L-lactate pentahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)