Merck
  • Vicenin 2 isolated from Artemisia capillaris exhibited potent anti-glycation properties.
Vicenin 2 isolated from Artemisia capillaris exhibited potent anti-glycation properties.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-10)
Md Nurul Islam, Ishrat Jahan Ishita, Hyun Ah Jung, Jae Sue Choi
ABSTRACT
Vicenin 2, isolated from a traditionally used medicinal plant Artemisia capillaris, is a 6,8-di-C-glucoside of apigenin which has been previously reported to possess a wide variety of pharmacological activities including antioxidant, anti-inflammatory, anti-cancer, and hepatoprotective. However, there have not been any reports concerning its anti-diabetic potential until now. Therefore, in the present study, we evaluated the anti-diabetic potential of vicenin 2 via α-glucosidase, protein tyrosine phosphatase 1B (PTP1B), rat lens aldose reductase (RLAR), and advanced glycation end products (AGE) formation inhibitory assays. Vicenin 2 strongly inhibited α-glucosidase, PTP1B, and RLAR in the corresponding assays. In addition, vicenin 2 inhibited the formation of both fluorescent AGE and nonfluorescent AGE, e.g., CML, as well as the level of fructosamine in glucose-fructose-induced bovine serum albumin (BSA) glycation. In the test system, vicenin 2 suppressed glycation-induced protein oxidation by attenuating the formation of protein carbonyl groups as well as by inhibiting the modification of protein thiol groups. Moreover, vicenin 2 was found to be a potent inhibitor of glycation-induced formation of amyloid cross-β structures in BSA. Taken together, vicenin 2 might be a useful lead for the development of multiple target-oriented therapeutic modalities for the treatment of diabetes and diabetes-associated complications.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Aminoguanidine hydrochloride, ≥98%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
DL-Glyceraldehyde, dimer, 95%
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium azide, BioXtra
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, JIS special grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%